Learn More
Aviva Systems Biology Corp FAM71C Antibody - N-terminal region (ARP53179_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53179_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM71C. It was validated on Western Blot. . Alternate names: MGC39520ApplicationWB. Species reactivity: Human
Catalog No. 50-285-804