Learn More
Aviva Systems Biology Corp FAM71D Antibody - middle region (ARP41172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM71D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM71D. Alternate names: C14orf54, FLJ36130ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-57748