Learn More
Aviva Systems Biology Corp FAM78A Antibody - C-terminal region (ARP55411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM78A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FAM78A. Alternate names: C9orf59, FLJ00024, RP11-544A12.6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-032