Learn More
Aviva Systems Biology Corp FAM79B Antibody - N-terminal region (ARP55877_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55877_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM79B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM79B. Alternate names: FLJ41238, FLJ43694, MGC126599, MGC126601, FAM79BApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-61645