Learn More
Aviva Systems Biology Corp FAM80A Antibody - middle region (ARP55677_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55677_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM80A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM80A. Alternate names: MGC47816, RP11-157D18.1, NAAGS, FAM80A, NAAGS-IIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61575