Learn More
Aviva Systems Biology Corp FAM82A Antibody - middle region (ARP52752_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52752_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM82A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM82A. Alternate names: BLOCK18, FLJ32954, FLJ38143, MGC33318, PRO34163, PYST9371, RMD2, RMD4, RMD-2, FAM82AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60820