Learn More
Aviva Systems Biology Corp FAM82B Antibody - N-terminal region (ARP46236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM82B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM82B. Alternate names: CGI-90, FLJ20665, RMD1, RMD-1, FAM82BApplicationWB. Species reactivity: Human
Catalog No. 50-014-59282