Learn More
Aviva Systems Biology Corp FAM82C Antibody - middle region (ARP47400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM82C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM82C. Alternate names: FLJ10579, ptpip51, RMD3, RMD-3, FAM82CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59541