Learn More
Biomatik Corporation Fam83D Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11521
Encompass_Preferred
Fam83D Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to FAM83D. The Fam83D Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Protein Fam83D Protein (339-470Aa). Applications: ELISA, WB, IF. Target Synonyms: FAM83D. Accession Number: Q9H4H8. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9242