Learn More
Aviva Systems Biology Corp FAM83E Antibody - middle region (ARP57016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM83E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FAM83E. Alternate names: FLJ20200, MGC138175, MGC138177ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-53947