Learn More
Aviva Systems Biology Corp FAM84A Antibody - N-terminal region (ARP52806_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52806_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM84A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM84A. Alternate names: FLJ35392, NSE1, PP11517ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60838