Learn More
Aviva Systems Biology Corp FAM90A1 Antibody - N-terminal region (ARP57109_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57109_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM90A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM90A1. Alternate names: FLJ10408ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62082