Learn More
Aviva Systems Biology Corp FAM98B Antibody - N-terminal region (ARP55663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FAM98B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FAM98B. Alternate names: FLJ38426ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61569