Learn More
Medchemexpress LLC Famciclovir | 104227-87-4 | ≥98% | 321.33 | 500 MG
SDP
Catalog No. 5000386478
Encompass_Preferred
Famciclovir (BRL 42810) is an orally active nucleoside analogue and an antiviral agent with potent activities against HBV, HSV and VZV. It can be used for the research of herpesvirus infection. It effectively cures necrotic hepatitis in mice.
  • Orally active nucleoside analogue
  • Antiviral agent with potent activities against HBV, HSV and VZV
  • Used for the research of herpesvirus infection
Catalog No. 50-003-86478 Supplier Medchemexpress LLC Supplier No. HY17426500MG
Famciclovir (BRL 42810) is an orally active nucleoside analogue and an antiviral agent with potent activities against HBV, HSV and VZV. It can be used for the research of herpesvirus infection. It effectively cures necrotic hepatitis in mice.
  • Orally active nucleoside analogue
  • Antiviral agent with potent activities against HBV, HSV and VZV
  • Used for the research of herpesvirus infection