Learn More
Selleck Chemical LLC Famciclovir S2467-50mg
SDP
Catalog No. 503111663
Encompass_Preferred
Famciclovir (BRL-42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections
Catalog No. 50-311-1663 Supplier Selleck Chemical LLC Supplier No. S246750MG
Famciclovir (BRL-42810) is a guanine analogue antiviral drug used for the treatment of various herpesvirus infections