Learn More
Cayman Chemical Famciclovir, Purity: ≥98%, CAS: 104227-87-4, 250 mg
SDP
Catalog No. 502994846
Encompass_Preferred
An orally bioavailable prodrug form of penciclovir; oxidized in vitro by human, guinea pig, and rat liver aldehyde oxidase; in rats the peak concentrations of penciclovir in plasma (mean 3.5 μg/ml) occur in 0.5 hours (40 mg/kg); in dogs the peak concentrations of penciclovir in plasma (mean 4.4 μg/ml) occur in 3 hours (25 mg/kg)
Catalog No. 50-299-4846 Supplier Cayman Chemical Supplier No. 23834250 MG
An orally bioavailable prodrug form of penciclovir; oxidized in vitro by human, guinea pig, and rat liver aldehyde oxidase; in rats the peak concentrations of penciclovir in plasma (mean 3.5 μg/ml) occur in 0.5 hours (40 mg/kg); in dogs the peak concentrations of penciclovir in plasma (mean 4.4 μg/ml) occur in 3 hours (25 mg/kg)