Learn More
Cayman Chemical FamcIclovIr 25mg
SDP
Catalog No. 502993680
Encompass_Preferred
An orally bioavailable prodrug form of penciclovir; oxidized in vitro by human, guinea pig, and rat liver aldehyde oxidase; in rats the peak concentrations of penciclovir in plasma (mean 3.5 μg/ml) occur in 0.5 hours (40 mg/kg); in dogs the peak concentrations of penciclovir in plasma (mean 4.4 μg/ml) occur in 3 hours (25 mg/kg)
Catalog No. 50-299-3680 Supplier Cayman Chemical Supplier No. 2383425 MG
An orally bioavailable prodrug form of penciclovir; oxidized in vitro by human, guinea pig, and rat liver aldehyde oxidase; in rats the peak concentrations of penciclovir in plasma (mean 3.5 μg/ml) occur in 0.5 hours (40 mg/kg); in dogs the peak concentrations of penciclovir in plasma (mean 4.4 μg/ml) occur in 3 hours (25 mg/kg)