Learn More
TARGETMOL CHEMICALS INC Famciclovir 500MG
SDP
Catalog No. 502876075
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Famciclovir (BRL 42810) is a Herpes Simplex Virus Nucleoside Analog DNA Polymerase Inhibitor. The mechanism of action of famciclovir is as a DNA Polymerase Inhibitor, and DNA Polymerase Inhibitor. Purity 99.58%
Catalog No. 50-287-6075 Supplier TARGETMOL CHEMICALS INC Supplier No. T1646500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Famciclovir (BRL 42810) is a Herpes Simplex Virus Nucleoside Analog DNA Polymerase Inhibitor. The mechanism of action of famciclovir is as a DNA Polymerase Inhibitor, and DNA Polymerase Inhibitor. Purity 99.58%