Learn More
Clinton Industries FAMILY PRACTICE EXAM TABLE
Catalog No. NC0108984
Encompass_Preferred
•Premium, vinyl upholstery over 2" firm, foam padding (5 cm) •Full 28.5" width for patient comfort (72.39 cm) •Air-spring adjustable backrest •Coated, two position adjustable stirrups •Pull-out leg rest on steel track •Optional pull-out basin •Easy care all laminate base •Color coordinated doors and top available •Chip resistant edges •1 large side storage compartment with shelf •Storage compartment is accessible from both sides (no right or left handed tables) •2 drawers with all steel slides •Built with plywood and laminate panels that are California CARB II compliant •Top sections are easily replaceable in the field •Adjustable feet for leveling •Paper dispenser included •400 lbs. load capacity under normal use (181.43 kg)
Catalog No. NC0108984 Supplier Clinton Industries Supplier No. 8870
Add to Cart
Edge
Add to Cart
•Premium, vinyl upholstery over 2" firm, foam padding (5 cm) •Full 28.5" width for patient comfort (72.39 cm) •Air-spring adjustable backrest •Coated, two position adjustable stirrups •Pull-out leg rest on steel track •Optional pull-out basin •Easy care all laminate base •Color coordinated doors and top available •Chip resistant edges •1 large side storage compartment with shelf •Storage compartment is accessible from both sides (no right or left handed tables) •2 drawers with all steel slides •Built with plywood and laminate panels that are California CARB II compliant •Top sections are easily replaceable in the field •Adjustable feet for leveling •Paper dispenser included •400 lbs. load capacity under normal use (181.43 kg)