Learn More
MSC SC Johnson Professional Disinfectant Spray, Fresh Scent, 17.5 oz, 8/pack
Catalog No. 502836606
Encompass
This multi-purpose aerosol disinfectant spray is designed to disinfect and deodorize hard, non-porous surfaces. It is also effective at neutralizing odor-causing bacteria on soft and sensitive surfaces. This spray is available in a fresh scent and comes in a case of eight 17.5 oz cans.
  • Kills 99.9% of germs and viruses on hard, non-porous surfaces.
  • Disinfects over 100 hard, non-porous surfaces.
  • Neutralizes odor-causing bacteria on soft and sensitive surfaces.
  • Sanitizes hard, non-porous surfaces in 30 seconds.
  • Effective against Covid-19 and flu viruses on hard, non-porous surfaces.
  • Inhibits mold and mildew on hard, non-porous surfaces.
Catalog No. 50-283-6606 Supplier MSC Supplier No. 48756787
This multi-purpose aerosol disinfectant spray is designed to disinfect and deodorize hard, non-porous surfaces. It is also effective at neutralizing odor-causing bacteria on soft and sensitive surfaces. This spray is available in a fresh scent and comes in a case of eight 17.5 oz cans.
  • Kills 99.9% of germs and viruses on hard, non-porous surfaces.
  • Disinfects over 100 hard, non-porous surfaces.
  • Neutralizes odor-causing bacteria on soft and sensitive surfaces.
  • Sanitizes hard, non-porous surfaces in 30 seconds.
  • Effective against Covid-19 and flu viruses on hard, non-porous surfaces.
  • Inhibits mold and mildew on hard, non-porous surfaces.