Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Famotidine, 98+%

Catalog No. AAJ6316506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6316506 5 g
AAJ6316503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6316506 Supplier Thermo Scientific Chemicals Supplier No. J6316506
Add to Cart

missing translation for 'validMainframeMsg'

A competitive histamine H2 receptor antagonist | CAS: 76824-35-6 | C8H15N7O2S3 | 337.435 g/mol

Famotidine, is commonly used in the treatment of peptic ulcer disease and gastroesophageal reflux disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and methanol

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76824-35-6
Molecular Formula C8H15N7O2S3
Molecular Weight (g/mol) 337.435
MDL Number MFCD00079297
InChI Key XUFQPHANEAPEMJ-UHFFFAOYSA-N
Synonym famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
PubChem CID 5702160
ChEBI CHEBI:4975
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
SMILES C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N

Specifications

Melting Point 166°C to 168°C
Color White
Quantity 5 g
Merck Index 14,3930
Solubility Information Soluble in water and methanol
Formula Weight 337.45
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Famotidine
RTECSNumber UA2300000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.