Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Famotidine, MP Biomedicals™

Catalog No. ICN15802394 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15802394 5 g
ICN15802390 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15802394 Supplier MP Biomedicals, Inc Supplier No. 0215802394

Chemical Identifiers

CAS 76824-35-6
Molecular Formula C8H15N7O2S3
Molecular Weight (g/mol) 337.435
InChI Key XUFQPHANEAPEMJ-UHFFFAOYSA-N
Synonym famotidine, pepcid ac, apogastine, amfamox, antodine, bestidine, digervin, dispromil, famodil, gastridin
PubChem CID 5702160
ChEBI CHEBI:4975
IUPAC Name 3-[[2-(diaminomethylideneamino)-1,3-thiazol-4-yl]methylsulfanyl]-N'-sulfamoylpropanimidamide
SMILES C1=C(N=C(S1)N=C(N)N)CSCCC(=NS(=O)(=O)N)N

Specifications

Color White
Quantity 5 g
Formula Weight 337.43
Physical Form Granular Powder
Chemical Name or Material Famotidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.