Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Famotidine, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18602598 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-598 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-602-598 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F11545GM

F1154, 76824-35-6, C8H15N7O2S3

Spectrum™ Chemical Famotidine, USP is a histamine and H2 receptor antagonist used to treat peptic ulcers and heartburn. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 76824-35-6
Molecular Formula C8H15N7O2S3
Molecular Weight (g/mol) 337.44
InChI Key XUFQPHANEAPEMJ-UHFFFAOYSA-N
IUPAC Name 3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
SMILES NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98.5 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.