Learn More
Sheldon Manufacturing Inc FAN ASSEMBLY
SDP
Catalog No. NC0631004
Encompass_Preferred
Blower motor assembly for several models of laboratory incubator manufactured by Sheldon Manufacturing. Contains motor, blower fan, and motor mount. Please contact Sheldon Manufacturing technical services to verify the parts you neeed for service purposes.
Catalog No. NC0631004 Supplier Sheldon Manufacturing Inc Supplier No. 9600593
Blower motor assembly for several models of laboratory incubator manufactured by Sheldon Manufacturing. Contains motor, blower fan, and motor mount. Please contact Sheldon Manufacturing technical services to verify the parts you neeed for service purposes.