Learn More
Vernier™ Fan Cart
SDPSCI_ED
Catalog No. S16111ND Shop All Vernier Software Products
Change view
Click to view available options
Includes:
Mass bars, sail, user manual
Catalog No. Includes
S16111ND Mass bars, sail, user manual
1 options
Catalog No. S16111ND Supplier Vernier™ Supplier No. CARTF
Add to Cart
Add to Cart
Includes
Fan Cart, Mass bars (2), Sail, User manual, The Fan Cart requires four AA batteries (not included).
Requires
For use with Vernier Dynamics System
The Fan Cart is a large fan on a light-weight cart. It offers students the ability to perform kinematics and dynamics experiments with constant acceleration, variable mass, variable thrust, and variable thrust angle.
  • Large fan on a light-weight cart
  • The fan has three speeds for studying the effect of variable thrust. Two mass bars let you change the total mass of the cart to observe how mass affects acceleration. The fan turns on a protractor base, allowing the thrust to be directed at a known angle from the direction of travel. In this way students can investigate vector component applications.
  • A sail is included to perform the fan-on-a-sailboat experiment. The sail can be mounted in two positions to study the subtle effects of diverted airflow
  • Although the cart can be used for qualitative experiments alone, most often it is used with a Motion Detector on a Dynamics Track.
Warranty and Services
5-year limited warranty
Specifications
Product Type Cart
For Use With (Application) Educational use only
For Use With (Equipment) Dynamics Cart and Track System, Dynamics System
Includes Mass bars, sail, user manual