Learn More
Newcomer Supply FAN PAD-GL Elite (Formalin Absorbing & Neutralizing Pad), 11" x 15", 100/case, Excellent for Grossing!
SDP
Catalog No. NC1318436
Encompass
The FAN Pad-GL Elite ™ (Formalin Absorbing Neutralizing Pad with Laminate Coating) absorbs and neutralizes formalin, destroying harmful vapors while you are dissecting tissue. The permeable non-woven blue laminate work surface gives excellent tissue contrast. The non-stick surface has exceptional knife suppression while dissecting tissue and is soft enough to reduce wear on dissecting blades. Can be used for: Specimen transport carts, boxes and coolers, Specimen/Biopsy cassette transfers, Surgery, Labor & Delivery, Specimen storage areas, Autopsy and GI Lab & Endoscopy. Dimensions: 11" x 15".
Catalog No. NC1318436 Supplier Newcomer Supply Supplier No. ABFAN1115E
The FAN Pad-GL Elite ™ (Formalin Absorbing Neutralizing Pad with Laminate Coating) absorbs and neutralizes formalin, destroying harmful vapors while you are dissecting tissue. The permeable non-woven blue laminate work surface gives excellent tissue contrast. The non-stick surface has exceptional knife suppression while dissecting tissue and is soft enough to reduce wear on dissecting blades. Can be used for: Specimen transport carts, boxes and coolers, Specimen/Biopsy cassette transfers, Surgery, Labor & Delivery, Specimen storage areas, Autopsy and GI Lab & Endoscopy. Dimensions: 11" x 15".