Learn More
Newcomer Supply FAN PAD-GL Elite (Formalin Absorbing & Neutralizing Pad), 8" x 11", 100/case, Excellent for Grossing!
SDP
Catalog No. NC1318435
Encompass
The FAN Pad-GL Elite ™ (Formalin Absorbing Neutralizing Pad with Laminate Coating) absorbs and neutralizes formalin, destroying harmful vapors while you are dissecting tissue. The permeable non-woven blue laminate work surface gives excellent tissue contrast. The non-stick surface has exceptional knife suppression while dissecting tissue and is soft enough to reduce wear on dissecting blades. Can be used for: Specimen transport carts, boxes and coolers, Specimen/Biopsy cassette transfers, Surgery, Labor & Delivery, Specimen storage areas, Autopsy and GI Lab & Endoscopy. Dimensions: 8" x 11".
Catalog No. NC1318435 Supplier Newcomer Supply Supplier No. ABFAN811E
The FAN Pad-GL Elite ™ (Formalin Absorbing Neutralizing Pad with Laminate Coating) absorbs and neutralizes formalin, destroying harmful vapors while you are dissecting tissue. The permeable non-woven blue laminate work surface gives excellent tissue contrast. The non-stick surface has exceptional knife suppression while dissecting tissue and is soft enough to reduce wear on dissecting blades. Can be used for: Specimen transport carts, boxes and coolers, Specimen/Biopsy cassette transfers, Surgery, Labor & Delivery, Specimen storage areas, Autopsy and GI Lab & Endoscopy. Dimensions: 8" x 11".