Learn More
StatLab Fan Pad-GL Formalin Neutralizing Pads
SDP
Catalog No. 22000036
Change view
Click to view available options
Size:
Large
Mini
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-000-036 Large
22-000-035 Mini
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-036 Supplier StatLab Supplier No. FAN825
Add to Cart
Add to Cart
Specially treated pad that absorbs formaldehyde
  • After absorption, formaldehyde is converted to a non-hazardous polymer eliminating harmful vapors
  • Fan Pad minis are also especially useful in the clean up of small formalin spills
Specifications
Material Polymer
Type Absorbent Pad
Disposable Limited-use
Size Large
Weight Capacity Heavyweight