Learn More
Newcomer Supply FAN PAD-GL Small (Formalin Absorbing & Neutralizing Pad), 6" x 14", 48/case
SDP
Catalog No. NC1084502
Encompass
The FAN Pads (Formalin Absorbing Neutralizing Pads)are an excellent product for cleaning up spills, grossing on and using as a shelf liner where formalin specimens are stored or transported. The Small FAN Pad is ideal for grossing medium size tissues (e.g. gall bladders and appendix). It absorbs up to 200ml of 10% Formalin. Working Surface: 6" x 14".
Catalog No. NC1084502 Supplier Newcomer Supply Supplier No. ABFAN614
The FAN Pads (Formalin Absorbing Neutralizing Pads)are an excellent product for cleaning up spills, grossing on and using as a shelf liner where formalin specimens are stored or transported. The Small FAN Pad is ideal for grossing medium size tissues (e.g. gall bladders and appendix). It absorbs up to 200ml of 10% Formalin. Working Surface: 6" x 14".