Learn More
World Precision Instrument Fan Track Stand with 76mm mount
SDP
Catalog No. 502521354
Encompass
This post stand comes with a 95mm plastic plate that is white on one side and black on the other. It is perfect for any microscope requiring a 76mm focus mount.
Catalog No. 50-252-1354 Supplier World Precision Instrument Supplier No. 502002
This post stand comes with a 95mm plastic plate that is white on one side and black on the other. It is perfect for any microscope requiring a 76mm focus mount.