Learn More
TARGETMOL CHEMICALS INC FANANSERIN 5MG
SDP
Catalog No. 502581238
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fananserin is an effective selective and oral active antagonist of 5-HT2 (Ki = 0.37 nM for the rat 5-HT2A). Fananserin is an antagonist of human D4 receptor (Ki = 2.93 nM). purity: 98%
Catalog No. 50-258-1238 Supplier TARGETMOL CHEMICALS INC Supplier No. T152705MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fananserin is an effective selective and oral active antagonist of 5-HT2 (Ki = 0.37 nM for the rat 5-HT2A). Fananserin is an antagonist of human D4 receptor (Ki = 2.93 nM). purity: 98%