Learn More
Medchemexpress LLC HY-15069A 10mg Medchemexpress, Fanapanel hydrate CAS:1255517-78-2 Purity:>98%
SDP
Catalog No. 501932722
Encompass_Preferred
Medchemexpress, HY-15069A 10mg Fanapanel hydrate CAS:1255517-78-2 Fanapanel hydrate (ZK200775 hydrate) is a highly selective AMPA/kainate antagonist with little activity against NMDA; have Ki values of 3.2 nM, 100 nM, and 8.5 μM against quisqualate, kainate, and NMDA, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2722 Supplier Medchemexpress LLC Supplier No. HY15069A10MG
Medchemexpress, HY-15069A 10mg Fanapanel hydrate CAS:1255517-78-2 Fanapanel hydrate (ZK200775 hydrate) is a highly selective AMPA/kainate antagonist with little activity against NMDA; have Ki values of 3.2 nM, 100 nM, and 8.5 μM against quisqualate, kainate, and NMDA, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.