Learn More
Aviva Systems Biology Corp FANCA Antibody - N-terminal region (ARP41678_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41678_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FANCA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FANCA. Alternate names: FA, FA-H, FA1, FAA, FACA, FAH, FANCH, MGC75158ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-014-57946