Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FANCC Human His-SU 10ug
SDP

Catalog No. 5000183247
Encompass_Preferred

Recombinant full-length human FANCC protein (residues M1-V558) with an N-terminal 6xHis-SUMO tag, expressed in E. coli and supplied lyophilized at 10 μg for research use. The product is intended for studies of DNA repair and Fanconi anemia pathways and includes specifications for purity, endotoxin, formulation, and storage.

  • Recombinant full-length human FANCC with N-terminal 6xHis and SUMO tag.
  • Supplied as a 10 μg lyophilized product suitable for biochemical assays.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 79.4 kDa.
  • Endotoxin level below 1 EU/μg (LAL method).
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.
  • Do not reconstitute below 100 μg/mL in ddH2O to avoid stability issues.

Catalog No. 50-001-83247 Supplier Medchemexpress LLC Supplier No. HYP7219010UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full-length human FANCC protein (residues M1-V558) with an N-terminal 6xHis-SUMO tag, expressed in E. coli and supplied lyophilized at 10 μg for research use. The product is intended for studies of DNA repair and Fanconi anemia pathways and includes specifications for purity, endotoxin, formulation, and storage.

  • Recombinant full-length human FANCC with N-terminal 6xHis and SUMO tag.
  • Supplied as a 10 μg lyophilized product suitable for biochemical assays.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight 79.4 kDa.
  • Endotoxin level below 1 EU/μg (LAL method).
  • Lyophilized from PBS with 6% trehalose, pH 7.4.
  • Store frozen; aliquot and keep at -20°C or -80°C for extended storage.
  • Do not reconstitute below 100 μg/mL in ddH2O to avoid stability issues.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.