Learn More
Medchemexpress LLC FANCC Human His-SU 50ug
SDP
Catalog No. 5000183360
Encompass_Preferred
Recombinant human FANCC protein (residues M1-V558) expressed with an N-terminal 6xHis-SUMO tag and supplied lyophilized for research use. The protein is >90% pure by reducing SDS-PAGE, has an approximate molecular weight of 79.4 kDa, and is formulated in PBS with 6% trehalose for enhanced stability. Follow the manufacturer datasheet for reconstitution and storage recommendations.
Catalog No. 50-001-83360 Supplier Medchemexpress LLC Supplier No. HYP7219050UG
Recombinant human FANCC protein (residues M1-V558) expressed with an N-terminal 6xHis-SUMO tag and supplied lyophilized for research use. The protein is >90% pure by reducing SDS-PAGE, has an approximate molecular weight of 79.4 kDa, and is formulated in PBS with 6% trehalose for enhanced stability. Follow the manufacturer datasheet for reconstitution and storage recommendations.