Learn More
Aviva Systems Biology Corp FANCE Antibody - middle region (ARP55381_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55381_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FANCE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FANCE. Alternate names: FACE, FAEApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-61486