Learn More
StatLab FanPad Small Formalin Neutralizing Pads
SDP
Catalog No. 22031875
Change view
Click to view available options
Packaging Quantity:
48/Cs.
Catalog No. Packaging Quantity
22-031-875 48/Cs.
1 options
Catalog No. 22-031-875 Supplier StatLab Supplier No. FAN614
Add to Cart
Add to Cart
FanPad Small Formalin Neutralizing Pads - Formaling Absorbant Pads
  • Specially treated pad that absorbs formaldehyde
  • After absorption, formaldehyde is converted to a non-hazardous polymer eliminating harmful vapors
  • Small 6 x 14 in. pads
  • 48 pads per case
Specifications
Type Formalin Neutralizer
Packaging Quantity 48/Cs.
Size Small
Dimensions (L x W) 6 x 14 in.