Learn More
MSC SC Johnson Professional Disinfectant Degreaser, 32 oz, Trigger, 1 Bottle
Catalog No. 502867971
Encompass
SC Johnson Professional fantastik Multi-Surface Disinfectant Degreaser is a multi-purpose daily cleaner that cleans, disinfects, degreases, and deodorizes surfaces, leaving a pleasant scent.
  • Removes 100% soap scum and grease
  • Kills 99.9% of viruses & bacteria when used as directed on hard non-porous surfaces
  • Bottle made of 100% post-consumer recycled plastic
  • Effective on a variety of surfaces and materials including stainless steel, sealed granite, and tile
  • EPA registered, Kosher and NSF certified
Catalog No. 50-286-7971 Supplier MSC Supplier No. 13356621
SC Johnson Professional fantastik Multi-Surface Disinfectant Degreaser is a multi-purpose daily cleaner that cleans, disinfects, degreases, and deodorizes surfaces, leaving a pleasant scent.
  • Removes 100% soap scum and grease
  • Kills 99.9% of viruses & bacteria when used as directed on hard non-porous surfaces
  • Bottle made of 100% post-consumer recycled plastic
  • Effective on a variety of surfaces and materials including stainless steel, sealed granite, and tile
  • EPA registered, Kosher and NSF certified