Learn More
MSC Fantech Direct Drive Fan, 7 in. Blade, 1 ea.
Catalog No. 5000506741
Encompass
The Fantech direct drive fan features a 7-inch blade and a 1/30 hp motor, providing a maximum airflow of 140 CFM. Designed for single-phase operation, it supports both vertical and horizontal mounting configurations.
  • Direct drive design
  • Vertical and horizontal mounting
  • Seven-inch blade
  • One-thirtieth hp motor
  • Maximum airflow of 140 CFM
  • Single phase operation
  • Galvanized steel construction
Catalog No. 50-005-06741 Supplier MSC Supplier No. 74077512
The Fantech direct drive fan features a 7-inch blade and a 1/30 hp motor, providing a maximum airflow of 140 CFM. Designed for single-phase operation, it supports both vertical and horizontal mounting configurations.
  • Direct drive design
  • Vertical and horizontal mounting
  • Seven-inch blade
  • One-thirtieth hp motor
  • Maximum airflow of 140 CFM
  • Single phase operation
  • Galvanized steel construction