Learn More
Medchemexpress LLC Fantofarone | 114432-13-2 | MFCD00867144 | 99.9% | 550.71 g·mol⁻¹ | C31H38N2O5S | 25 MG
SDP
Catalog No. 5000243497
Encompass_Preferred
Fantofarone is a potent L-type calcium channel antagonist used as a research reagent to investigate calcium channel pharmacology and related cardiovascular signaling pathways. It is a sulfone-indolizine class compound with molecular formula C31H38N2O5S and a molecular weight of 550.71 g·mol⁻¹. The material is supplied for research use only and is not intended for human or veterinary use.
  • Potent L-type calcium channel antagonist for in vitro pharmacology.
  • High reported purity suitable for analytical characterization.
  • Solid reagent convenient for formulation and dosing in laboratory assays.
  • Supplied in small research quantities appropriate for experimental workflows.
Catalog No. 50-002-43497 Supplier Medchemexpress LLC Supplier No. HY10511725MG
Fantofarone is a potent L-type calcium channel antagonist used as a research reagent to investigate calcium channel pharmacology and related cardiovascular signaling pathways. It is a sulfone-indolizine class compound with molecular formula C31H38N2O5S and a molecular weight of 550.71 g·mol⁻¹. The material is supplied for research use only and is not intended for human or veterinary use.
  • Potent L-type calcium channel antagonist for in vitro pharmacology.
  • High reported purity suitable for analytical characterization.
  • Solid reagent convenient for formulation and dosing in laboratory assays.
  • Supplied in small research quantities appropriate for experimental workflows.