Learn More
Medchemexpress LLC HY-147057 1mg , FAP-2286 CAS:2581741-18-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1470571MG
Encompass_Preferred
HY-147057 1mg FAP-2286 CAS: 2581741-18-4 FAP-2286, a fibroblast activation protein (FAP)-binding peptidic macrocycle coupled to the radionuclide chelator DOTA. FAP-2286 has potent affinity to human FAP protein with a Kd of 1.1 nM. FAP-2286, a tumor imaging agent, is a useful tool for the research of positron emission tomography (PET). FAP-2286 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1873