Learn More
BPS Bioscience Inc FAP Assay Kit, fluorogenic, 96 reactions, FAP detection, recombinant FAP enzyme, fluorogenic DPP substrate 1, DPP assay buffer, fluorescence detection, human species reactivity
SDP
Catalog No. NC1452553
Encompass
FAP Assay Kit, fluorogenic, 96 reactions, FAP detection, recombinant FAP enzyme, fluorogenic DPP substrate 1, DPP assay buffer, fluorescence detection, human species reactivity
Catalog No. NC1452553 Supplier BPS Bioscience Inc Supplier No. 80210
FAP Assay Kit, fluorogenic, 96 reactions, FAP detection, recombinant FAP enzyme, fluorogenic DPP substrate 1, DPP assay buffer, fluorescence detection, human species reactivity