Learn More
Enzo Life Sciences Fibroblast activation protein (human) monoclonal antibody (F11-24) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804327C100
Encompass_Preferred
Clone: F11-24. Isotype: IgG1. Immunogen: Human FAP (fibroblast activation protein). Species reactivity: Human. Applications: Flow Cytometry, IHC (FS), IP. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS.
Catalog No. 50-200-9866