Learn More
Enzo Life Sciences FAP (human), (recombinant) (His-tag) (10 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE4090010
Encompass_Preferred
Alternative name: Fibroblast activation protein alpha, Seprase, Surface expressed protease, APCE, Antiplasmin cleaving enzyme. Formulation: Liquid. In 40mM TRIS-HCl, pH 8.0, containing 110mM NaCl, 2.2mM KCl, 200mM imidazole, 20% glycerol and 3mM DTT. Source: Produced in a baculovirus expression system. Active recombinant human FAP (aa 29-760), representing a naturally-occurring cleaved (soluble) form fused at the C-terminus to a His-tag. Long Term Storage: -80°C.
Catalog No. 50-201-2013