Learn More
Medchemexpress LLC FAP Human Biotinyl 20ug | 20ug
SDP
Catalog No. 5000187571
Encompass_Preferred
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived FAP protein expressed by HEK293 with N-His N-Avi labeled tag
Catalog No. 50-001-87571 Supplier Medchemexpress LLC Supplier No. HYP7812120UG
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived FAP protein expressed by HEK293 with N-His N-Avi labeled tag