Learn More
Medchemexpress LLC FAP Human HEK293 250ug
SDP
Catalog No. 5000181411
Encompass_Preferred
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Human (HEK293 His) is the recombinant human-derived FAP protein expressed by HEK293 with N-8 His labeled tag
Catalog No. 50-001-81411 Supplier Medchemexpress LLC Supplier No. HYP72659A250UG
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Human (HEK293 His) is the recombinant human-derived FAP protein expressed by HEK293 with N-8 His labeled tag