Learn More
Medchemexpress LLC FAP Rat HEK293 Hi 50ug | 50ug
SDP
Catalog No. 5000194490
Encompass_Preferred
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Rat (HEK293 His) is the recombinant rat-derived FAP protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-94490 Supplier Medchemexpress LLC Supplier No. HYP7862250UG
The FAP protein is a cell surface glycoprotein serine protease that is critical for extracellular matrix degradation and plays multiple roles in tissue remodeling fibrosis wound healing inflammation and tumor growth FAP Protein Rat (HEK293 His) is the recombinant rat-derived FAP protein expressed by HEK293 with N-His labeled tag