Learn More
Medchemexpress LLC HY-137331 5mg , FAPI-46 CAS:2374782-04-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1373315MG
Encompass_Preferred
HY-137331 5mg FAPI-46 CAS: 2374782-04-2 FAPI-46 is a quinoline-based fibroblast activation protein (FAP)-targeted radiotracer. FAPI-46 has higher tumor uptake and prolonged tumor accumulation. FAPI-46 can be used for tumor imaging of a multitude of different cancers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9248