Learn More
Medchemexpress LLC HY-113303 10mg Medchemexpress, FAPy-adenine CAS:5122-36-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11330310MG
Encompass_Preferred
Medchemexpress, HY-113303 10mg FAPy-adenine CAS:5122-36-1 FAPy-adenine is an oxidized DNA base. Fapy-adenine shows an increased trend levels in the Alzheimer's disease brain. Oxidized nucleosides are biochemical markers for tumors, aging, and neurodegenerative diseases[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9759