Learn More
Medchemexpress LLC FAPy-adenine | 5122-36-1 | 99.86% | 153.14 | 1 ML
SDP
Catalog No. 5000436693
Encompass_Preferred
FAPy-adenine is an oxidized DNA base whose levels show an increased trend in the Alzheimer's disease brain. Oxidized nucleosides serve as biochemical markers for tumors, aging, and neurodegenerative diseases. This compound is able to form pairs with all four canonical nucleic acid bases. For research use only; not sold to patients.
  • Oxidized DNA base.
  • Increased levels observed in Alzheimer's disease brain.
  • Biochemical marker for tumors, aging, and neurodegenerative diseases.
  • Forms pairs with all four canonical nucleic acid bases.
Catalog No. 50-004-36693 Supplier Medchemexpress LLC Supplier No. HY11330310MM/1ML
FAPy-adenine is an oxidized DNA base whose levels show an increased trend in the Alzheimer's disease brain. Oxidized nucleosides serve as biochemical markers for tumors, aging, and neurodegenerative diseases. This compound is able to form pairs with all four canonical nucleic acid bases. For research use only; not sold to patients.
  • Oxidized DNA base.
  • Increased levels observed in Alzheimer's disease brain.
  • Biochemical marker for tumors, aging, and neurodegenerative diseases.
  • Forms pairs with all four canonical nucleic acid bases.